Learn More
IDEX Health & Science Upchurch ADAPTER CROSS 360, FOR 1/16 OD
Catalog No. NC1788156
Encompass_Preferred
VHP MicroTight Adapter Cross , 360, for 1/16" OD; Color: Black; Swept Volume uL: 0.11; Thread Port Configuration Coned: 10-32 to 5/16-24; Material: Stainless Steel; Pressure Rating Max (psi): 15000;Pressure Rating Max (bar): 1035; For Tubing OD (um):360. IDEX HEALTH & SCIENCE UPCHURCH mft# UH-906
Catalog No. NC1788156 Supplier IDEX Health & Science Upchurch Supplier No. UH906
VHP MicroTight Adapter Cross , 360, for 1/16" OD; Color: Black; Swept Volume uL: 0.11; Thread Port Configuration Coned: 10-32 to 5/16-24; Material: Stainless Steel; Pressure Rating Max (psi): 15000;Pressure Rating Max (bar): 1035; For Tubing OD (um):360. IDEX HEALTH & SCIENCE UPCHURCH mft# UH-906