Learn More
World Precision Instrument 13324 Adapter, Dual Banana/BNC
SDP
Catalog No. 50822008
Encompass
For wiring any laboratory setup
  • Connector A: Double-banana (female)
  • Connector B: BNC (male)
  • Cable: none
Catalog No. 50-822-008 Supplier World Precision Instrument Supplier No. 13324
For wiring any laboratory setup
  • Connector A: Double-banana (female)
  • Connector B: BNC (male)
  • Cable: none