Learn More
TA Instruments Adapter Discovery PCA, Mechanical Accessory for Proper Integration of PCA with Discovery DSC Instruments, Ensures Secure Mounting and Alignment
Catalog No. 5000580887 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Adapter Discovery PCA is a mechanical accessory designed to facilitate proper mounting and alignment of the PCA with Discovery DSC instruments. It ensures accurate positioning for optimal optical performance and reliable measurement results. Precision-engineered for compatibility, the adapter enhances ease of use while maintaining secure stability during experiments. This accessory is essential for laboratories utilizing Discovery DSC systems with PCA, ensuring seamless integration and consistent experimental performance.
Catalog No. 50-005-80887 Supplier TA Instruments Supplier No. 935050.001
Waters TA Instruments Adapter Discovery PCA is a mechanical accessory designed to facilitate proper mounting and alignment of the PCA with Discovery DSC instruments. It ensures accurate positioning for optimal optical performance and reliable measurement results. Precision-engineered for compatibility, the adapter enhances ease of use while maintaining secure stability during experiments. This accessory is essential for laboratories utilizing Discovery DSC systems with PCA, ensuring seamless integration and consistent experimental performance.