Learn More
Chemglass Life Sciences Adapter, Distillation, Claisen, 14/20 Joint Size, 10/18 Thermometer Joint For 25mm Immersion, 105mm H x 100mm W
SDP
Catalog No. CG101514
Encompass_Preferred
Adapter, Distillation, Claisen Joint Size, 10/18 Thermometer Joint For 25mm Immersion, 105mm H x 100mm W. Distilling adapter having an outer member on the vertical side tube for use with a thermometer of the listed joint size and immersion. Sidearm has a standard taper inner drip tip joint while vertical tube has a top outer and lower inner joint, all being the same size. FOR THERMOMETERS SEE CG-3506-N.
Catalog No. CG101514 Supplier Chemglass Life Sciences Supplier No. CG101514
Adapter, Distillation, Claisen Joint Size, 10/18 Thermometer Joint For 25mm Immersion, 105mm H x 100mm W. Distilling adapter having an outer member on the vertical side tube for use with a thermometer of the listed joint size and immersion. Sidearm has a standard taper inner drip tip joint while vertical tube has a top outer and lower inner joint, all being the same size. FOR THERMOMETERS SEE CG-3506-N.