Learn More
Chemglass Life Sciences Adapter, Distillation, 14/20 Joint, 100mm Center to Center
SDP
Catalog No. CG1072B14
Encompass_Preferred
Adapter, Distillation, 14/20 Joint, 100mm Center to Center. With two standard taper inner joints, parallel, and at a 75º angle to the connecting tube. Adapter has a sealed in drip tip and serrated hose connection for vacuum source.
Catalog No. CG1072B14 Supplier Chemglass Life Sciences Supplier No. CG1072B14
Adapter, Distillation, 14/20 Joint, 100mm Center to Center. With two standard taper inner joints, parallel, and at a 75º angle to the connecting tube. Adapter has a sealed in drip tip and serrated hose connection for vacuum source.