Learn More
Chemglass Life Sciences Adapter, Distillation, 14/20 Joint Size, Distance Between Inner Joints/100mm
SDP
Catalog No. CG107214
Encompass_Preferred
Adapter, Distillation, 14/20 Joint Size, Distance Between Inner Joints/100mm. With two standard taper inner joints, parallel, and at a 75º angle to the connecting tube.
Catalog No. CG107214 Supplier Chemglass Life Sciences Supplier No. CG107214
Adapter, Distillation, 14/20 Joint Size, Distance Between Inner Joints/100mm. With two standard taper inner joints, parallel, and at a 75º angle to the connecting tube.