Learn More
Chemglass Life Sciences Adapter, Distillation, 14/20 Inner Joints, Bent 75°
SDP
Catalog No. CG101014
Encompass_Preferred
Adapter, Distillation, 14/20 Inner Joints, Bent 75¿ with standard taper inner joints at a 75¿ angle to one another.
Catalog No. CG101014 Supplier Chemglass Life Sciences Supplier No. CG101014
Adapter, Distillation, 14/20 Inner Joints, Bent 75¿ with standard taper inner joints at a 75¿ angle to one another.