Learn More
Chemglass Life Sciences Adapter, Distillation, 105°, 19/22 Joint Size
SDP
Catalog No. CG1071A19
Encompass_Preferred
Adapter, Distillation, 105¿, 19/22 Joint Size. Distillation adapter with sealed in lower drip tip and top outer joints, both being the same size.
Catalog No. CG1071A19 Supplier Chemglass Life Sciences Supplier No. CG1071A19
Adapter, Distillation, 105¿, 19/22 Joint Size. Distillation adapter with sealed in lower drip tip and top outer joints, both being the same size.