Learn More
Chemglass Life Sciences Adapter, Distillation, 105°, 19/22 Outer
SDP
Catalog No. CG107019
Encompass_Preferred
Adapter, Distillation, 105¿, 19/22 Outer. Distillation adapter with a top outer standard taper joint. Tapered delivery tube is bent at a 105¿ angle to the outer joint and extends into open receivers.
Catalog No. CG107019 Supplier Chemglass Life Sciences Supplier No. CG107019
Adapter, Distillation, 105¿, 19/22 Outer. Distillation adapter with a top outer standard taper joint. Tapered delivery tube is bent at a 105¿ angle to the outer joint and extends into open receivers.