Learn More
Chemglass Life Sciences Adapter, Distillation, 105°, 14/20 Joint Size
SDP
Catalog No. 501531001
Encompass_Preferred
Adapter, Distillation, 105, 14/20 Joint Size. Distillation adapter with sealed in lower drip tip and top outer joints, both being the same size.
Catalog No. 50-153-1001 Supplier Chemglass Life Sciences Supplier No. CG1071A14
Adapter, Distillation, 105, 14/20 Joint Size. Distillation adapter with sealed in lower drip tip and top outer joints, both being the same size.