Learn More
Chemglass Life Sciences Adapter, Distillation, 75°, 24/40 Inner Joints, #7 Chem-Thread
SDP
Supplier:  Chemglass Life Sciences CG102701
Encompass_Preferred
Adapter, Distillation, 75¿, 24/40 Inner Joints, #7 Chem-Thread. Adapter has a #7 Chem-Thread at the top for vacuum tight seal of plain stem thermometers. With the same size inner joints on the bottom and sidearm at a 75¿ angle to one another. Supplied complete with a compression cap and Viton o-ring. FOR THERMOMETERS SEE CG-3503.
Catalog No. 50-872-842