Learn More
Chemglass Life Sciences Adapter, Distillation, 24/40 Joint Size, Distance Between Inner Joints/200mm
SDP
Catalog No. 50872916
Encompass_Preferred
Adapter, Distillation, 24/40 Joint Size, Distance Between Inner Joints/200mm. With two standard taper inner joints, parallel, and at a 75º angle to the connecting tube.
Catalog No. 50-872-916 Supplier Chemglass Life Sciences Supplier No. CG107201
Adapter, Distillation, 24/40 Joint Size, Distance Between Inner Joints/200mm. With two standard taper inner joints, parallel, and at a 75º angle to the connecting tube.