Learn More
Chemglass Life Sciences Adapter, Distillation, 105°, 24/40 Joint Size
SDP
Catalog No. CG107101
Encompass_Preferred
Adapter, Distillation, 105¿, 24/40 Joint Size. Distillation adapter with top outer and lower inner joints both being the same size.
Catalog No. CG107101 Supplier Chemglass Life Sciences Supplier No. CG107101
Adapter, Distillation, 105¿, 24/40 Joint Size. Distillation adapter with top outer and lower inner joints both being the same size.