Learn More
Chemglass Life Sciences Adapter, Distillation, 29/42 Inner Joints, Bent 75°
SDP
Catalog No. CG101002
Encompass_Preferred
Adapter, Distillation, 29/42 Inner Joints, Bent 75¿, with standard taper inner joints at a 75¿ angle to one another.
Catalog No. CG101002 Supplier Chemglass Life Sciences Supplier No. CG101002
Adapter, Distillation, 29/42 Inner Joints, Bent 75¿, with standard taper inner joints at a 75¿ angle to one another.