Learn More
Chemglass Life Sciences Chemglass Adapter, Distillation, Bent - 45/50 Inner Joint to 65/40 O-Ring Ball Joint; for diagonal distillation cart systems (30/50 L reactors)
SDP
Catalog No. 502733552
Encompass_Preferred
Adapter used to connect a 45/50 inner ground joint to a 65/40 O-ring ball joint in distillation assemblies. It features a bent geometry to route vapor and provide a sealed transition for diagonal distillation cart systems and distillation kits used with 30 and 50 L process reactors.
  • Bent geometry routes vapor between mismatched joint types.
  • Connects a 45/50 inner ground joint to a 65/40 O-ring ball joint.
  • Designed for use in diagonal distillation cart systems and distillation kits for 30 and 50 L process reactors.
  • Manufactured glassware construction suitable for laboratory distillation assemblies.
  • Facilitates sealed transition and alignment in reactor distillation setups.
Catalog No. 50-273-3552 Supplier Chemglass Life Sciences Supplier No. CG1968D304
Adapter used to connect a 45/50 inner ground joint to a 65/40 O-ring ball joint in distillation assemblies. It features a bent geometry to route vapor and provide a sealed transition for diagonal distillation cart systems and distillation kits used with 30 and 50 L process reactors.
  • Bent geometry routes vapor between mismatched joint types.
  • Connects a 45/50 inner ground joint to a 65/40 O-ring ball joint.
  • Designed for use in diagonal distillation cart systems and distillation kits for 30 and 50 L process reactors.
  • Manufactured glassware construction suitable for laboratory distillation assemblies.
  • Facilitates sealed transition and alignment in reactor distillation setups.