Learn More
Chemglass Life Sciences Adapter, Distilling Trap, Kjeldahl, 29/42 Top Outer, 14/20 Lower Inner
SDP
Catalog No. CG1032A05
Encompass_Preferred
Adapter, Distilling Trap, Kjeldahl, 29/42 Top Outer, 14/20 Lower Inner. Similar to CG-1032, but with joints of non-similar sizes at the upper and lower portion of the adapter. Dimensions are the same as the CG-1032.
Catalog No. CG1032A05 Supplier Chemglass Life Sciences Supplier No. CG1032A05
Adapter, Distilling Trap, Kjeldahl, 29/42 Top Outer, 14/20 Lower Inner. Similar to CG-1032, but with joints of non-similar sizes at the upper and lower portion of the adapter. Dimensions are the same as the CG-1032.