Learn More
Chemglass Life Sciences Adapter, Distilling/Splitting, 24/40, 0-4mm Valve, 1/2" Compression Fitting, Process Reactor
SDP
Catalog No. 50143643
Encompass_Preferred
Adapter, Distilling/Splitting, 24/40, 0-4mm Valve, 1/2" Compression Fitting, Process Reactor. Compact and simple distilling/splitter adapter for use with Chemglass process reactors. Vapor flows upward freely through the large vapors holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor. Control valve is either a 0-4mm or a 0-12mm PTFE with a 1/2¿ or 3/4¿ OD distillate takeoff for use with CG-1975 compression fittings and TFE tubing. Plugs are supplied complete with CG-309 Perfluoro o-rings.
Catalog No. 50-143-643 Supplier Chemglass Life Sciences Supplier No. CG1235A24
Adapter, Distilling/Splitting, 24/40, 0-4mm Valve, 1/2" Compression Fitting, Process Reactor. Compact and simple distilling/splitter adapter for use with Chemglass process reactors. Vapor flows upward freely through the large vapors holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor. Control valve is either a 0-4mm or a 0-12mm PTFE with a 1/2¿ or 3/4¿ OD distillate takeoff for use with CG-1975 compression fittings and TFE tubing. Plugs are supplied complete with CG-309 Perfluoro o-rings.