Learn More
Wilmad Labglass Distilling Adapter with 75 Degree Angle Side Arm, Standard Taper 29/42
Catalog No. 50807380
Encompass_Preferred
With sidearm at angle of 75 from lower part of main tube. Sidearm and lower tube have inner joints; upper tube outer joint. All three joints are the same size.
Catalog No. 50-807-380 Supplier Wilmad Labglass Supplier No. LG1470102
With sidearm at angle of 75 from lower part of main tube. Sidearm and lower tube have inner joints; upper tube outer joint. All three joints are the same size.