Learn More
Chemglass Life Sciences Adapter, Distillation, 75°, 29/42 Inner Joints, 10/30 Thermometer Joint For 75mm Immersion
SDP
Catalog No. CG102402
Encompass_Preferred
Adapter, Distillation, 75¿, 29/42 Inner Joints, 10/30 Thermometer Joint For 75mm Immersion. Adapter has a standard taper outer joint at the top of the main tube for use with jointed thermometers. The 10/18 size is used with 10/18, 25mm immersion thermometers while the 10/30 is for use with 10/30, 75mm immersion thermometers. With inner joints on the bottom and sidearm at a 75¿ angle to one another.
Catalog No. CG102402 Supplier Chemglass Life Sciences Supplier No. CG102402
Adapter, Distillation, 75¿, 29/42 Inner Joints, 10/30 Thermometer Joint For 75mm Immersion. Adapter has a standard taper outer joint at the top of the main tube for use with jointed thermometers. The 10/18 size is used with 10/18, 25mm immersion thermometers while the 10/30 is for use with 10/30, 75mm immersion thermometers. With inner joints on the bottom and sidearm at a 75¿ angle to one another.