Learn More
Ace Glass, Inc. Adapter, distillation, (2)45/50 upper & (1)45/50 lower joints, 3/4in Swagelok stopcock fitting
SDP
Catalog No. 50971838
Encompass_Preferred
ADAPTER DISTILLING (3)45/50
Catalog No. 50-971-838 Supplier Ace Glass, Inc. Supplier No. 529916
ADAPTER DISTILLING (3)45/50