Learn More
Chemglass Life Sciences Adapter, Distilling Trap, Kjeldahl, 29/42 Joint Size
SDP
Catalog No. 50872849
Encompass_Preferred
Adapter, Distilling Trap, Kjeldahl, 29/42 Joint Size. Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam. With top standard taper outer and lower inner joints, both being the same size.
Catalog No. 50-872-849 Supplier Chemglass Life Sciences Supplier No. CG103202
Adapter, Distilling Trap, Kjeldahl, 29/42 Joint Size. Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam. With top standard taper outer and lower inner joints, both being the same size.