Learn More
Chemglass Life Sciences Adapter, Distillation, 24/40 - 10/30, 75°, Jacketed
SDP
Catalog No. CG107501
Encompass_Preferred
Adapter, Distillation, 24/40 - 10/30, 75¿, Jacketed. Adapter is vacuum jacketed and can be used as a distilling head to connect a condenser with the top of the distillation column. The 10/30 top outer joint is for use with CG-3506, 75mm immersion thermometers. Inner joints are at a 75¿ angle to one another and are both the same size. For thermometers see CG-3506-N.
Catalog No. CG107501 Supplier Chemglass Life Sciences Supplier No. CG107501
Adapter, Distillation, 24/40 - 10/30, 75¿, Jacketed. Adapter is vacuum jacketed and can be used as a distilling head to connect a condenser with the top of the distillation column. The 10/30 top outer joint is for use with CG-3506, 75mm immersion thermometers. Inner joints are at a 75¿ angle to one another and are both the same size. For thermometers see CG-3506-N.