Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Reducing adapter for DRB200, 20 to 16 mm

Catalog No. 501997623 Shop All Hach Company Products
Encompass

This adapter converts the 20 mm diameter vial wells to 16 mm diameter.

Catalog No. 50-199-7623 Supplier Hach Company Supplier No. HHA155
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This adapter converts the 20 mm diameter vial wells to 16 mm diameter.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.