Learn More
Perkin Elmer US LLC 1⁄4 in. (6.4 mm) Internal Screw Thread Adapter for the Quartz Cell, Type E
Catalog No. 509042333
Encompass_Preferred
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. This adapter is included in the Hydride Accessory Kit for Avio 200/500 (N0810433). Type E in instrument manual diagrams.
Catalog No. 50-904-2333 Supplier Perkin Elmer US LLC Supplier No. B0196857
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. This adapter is included in the Hydride Accessory Kit for Avio 200/500 (N0810433). Type E in instrument manual diagrams.