Learn More
Cole-Parmer Idex Threaded Luer Adapter, Natural Polypropylene, 0.050" Bore, Female Luer x Male 1/4-28 Flat Bottom; 1/EA
Supplier:  Cole-Parmer 0202338
Encompass
Convenience of quick connect and disconnect of a luer connectionManufactured from inert polymersFemale Luer x Male 1/4-28 Flat Bottom0.050" thru-hole diameterManufactured from natural polypropylene
Catalog No. NC2890720