Learn More
Chemglass Life Sciences Adapter, Vacuum Filtration, 23mm ID Flange For #2 Pluro Stopper, 14/20 Inner Joint, 51mm Overall Height
SDP
Catalog No. 50872887
Encompass_Preferred
Adapter, Vacuum Filtration, 23mm ID Flange For #2 Pluro Stopper, 14/20 Inner Joint, 51mm Overall Height. Adapter used for reduced pressure filtration with plain stem Buchner funnels. Top has a HEAVY DUTY tooled flare to accept the listed size pluro stopper. Hose connection is 10mm OD at the largest serration. Accepts up to 60mL plain stem Buchner Funnels. ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO STOPPER.
Catalog No. 50-872-887 Supplier Chemglass Life Sciences Supplier No. CG1052A14
Adapter, Vacuum Filtration, 23mm ID Flange For #2 Pluro Stopper, 14/20 Inner Joint, 51mm Overall Height. Adapter used for reduced pressure filtration with plain stem Buchner funnels. Top has a HEAVY DUTY tooled flare to accept the listed size pluro stopper. Hose connection is 10mm OD at the largest serration. Accepts up to 60mL plain stem Buchner Funnels. ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO STOPPER.