Learn More
Chemglass Life Sciences Adapter, Tooled Pluro Stopper Flare to 20-400 GPI Thread
SDP
Catalog No. 50872879
Encompass_Preferred
Adapter, Tooled Pluro Stopper Flare to 20-400 GPI Thread. Adapter with tooled flare top is used for reduced pressure filtration with plain stem Buchner funnels. Flare accepts a #2 Pluro Stopper. ADAPTER IS NOT SUPPLIED WITH PLURO STOPPER UNLESS PURCHASED AS A KIT. SEE CG-1401 FOR STOPPER. Adapters are used for filtration directly into vials. There are three top configurations, with the bottom of all adapters having a 20-400 GPI thread for connection to vials using a CG-1318 Threaded Connector. Design incorporates a drip tube sealed within thread to prevent breakage and are supplied with a 2-1/2” PTFE delivery stem. Hose connection is 10mm OD at the largest serration. ADAPTERS ARE SUPPLIED WITH PTFE DELIVERY STEM.
Catalog No. 50-872-879 Supplier Chemglass Life Sciences Supplier No. CG1049C02
Adapter, Tooled Pluro Stopper Flare to 20-400 GPI Thread. Adapter with tooled flare top is used for reduced pressure filtration with plain stem Buchner funnels. Flare accepts a #2 Pluro Stopper. ADAPTER IS NOT SUPPLIED WITH PLURO STOPPER UNLESS PURCHASED AS A KIT. SEE CG-1401 FOR STOPPER. Adapters are used for filtration directly into vials. There are three top configurations, with the bottom of all adapters having a 20-400 GPI thread for connection to vials using a CG-1318 Threaded Connector. Design incorporates a drip tube sealed within thread to prevent breakage and are supplied with a 2-1/2” PTFE delivery stem. Hose connection is 10mm OD at the largest serration. ADAPTERS ARE SUPPLIED WITH PTFE DELIVERY STEM.