Learn More
Chemrus Inc ADAPTER FIT 14/20 OUTER JOINT
Catalog No. 5082317
Encompass
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 14/20 outer joint
Catalog No. 50-823-17 Supplier Chemrus Inc Supplier No. CR202801
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 14/20 outer joint