Learn More
Chemrus Inc ADAPTER FIT 19/22 OUTER JOINT
Catalog No. 5082321
Encompass
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 19/22 outer joint
Catalog No. 50-823-21 Supplier Chemrus Inc Supplier No. CR202805
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 19/22 outer joint