Learn More
Chemrus Inc ADAPTER FIT 24/40 OUTER JOINT
Catalog No. 5082318
Encompass
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 24/40 outer joint
Catalog No. 50-823-18 Supplier Chemrus Inc Supplier No. CR202802
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 24/40 outer joint