Learn More
Chemrus Inc ADAPTER FIT 29/42 OUTER JOINT
Catalog No. 5082319
Encompass
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 29/42 outer joint
Catalog No. 50-823-19 Supplier Chemrus Inc Supplier No. CR202803
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 29/42 outer joint