Learn More
Beckman Coulter Adapter, 3.5mm to 5.5mm
Catalog No. NC0943793
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Rotor Adapter fits 0.2ml to 0.5ml tubes in the Microfuge 18 rotor. Made of polypropylene, it holds 18 tubes, reaches 13,300 rpm, and generates 16,060 x g. Dimensions are 3.9 x 3.9 x 1.4 inches, weight 0.32 lbs, and it's compatible with various Beckman Coulter centrifuges.
Catalog No. NC0943793 Supplier Beckman Coulter Supplier No. 355534
The Beckman Coulter Microfuge 18 Centrifuge Rotor Adapter fits 0.2ml to 0.5ml tubes in the Microfuge 18 rotor. Made of polypropylene, it holds 18 tubes, reaches 13,300 rpm, and generates 16,060 x g. Dimensions are 3.9 x 3.9 x 1.4 inches, weight 0.32 lbs, and it's compatible with various Beckman Coulter centrifuges.