Learn More
Chemglass Life Sciences Adapter, Flow Control, 2mm Stpk, 24/40 Joint Size
SDP
Catalog No. CG103101
Encompass_Preferred
Adapter, Flow Control, 2mm Stpk, 24/40 Joint Size. Adapter has a 2mm PTFE metering stopcock between top outer and lower inner standard taper joints to permit flow control of fluids between apparatus.
Catalog No. CG103101 Supplier Chemglass Life Sciences Supplier No. CG103101
Adapter, Flow Control, 2mm Stpk, 24/40 Joint Size. Adapter has a 2mm PTFE metering stopcock between top outer and lower inner standard taper joints to permit flow control of fluids between apparatus.