Learn More
Chemglass Life Sciences Adapter, Flow Control, 2mm Stpk, 14/20 Joint Size
SDP
Catalog No. 50872848
Encompass_Preferred
Adapter, Flow Control, 2mm Stpk, 14/20 Joint Size. Adapter has a 2mm PTFE metering stopcock between top outer and lower inner standard taper joints to permit flow control of fluids between apparatus.
Catalog No. 50-872-848 Supplier Chemglass Life Sciences Supplier No. CG103114
Adapter, Flow Control, 2mm Stpk, 14/20 Joint Size. Adapter has a 2mm PTFE metering stopcock between top outer and lower inner standard taper joints to permit flow control of fluids between apparatus.