Learn More
Chemglass Life Sciences Adapter, Flow Control, Straight, 2mm Stpk, 14/20 Inner Joint
SDP
Catalog No. 50872845
Encompass_Preferred
Adapter, Flow Control, Straight, 2mm Stpk, 14/20 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm and is supplied complete with a CG-423 retaining clip. Hose connection has an O.D. of 8mm at the middle serration.
Catalog No. 50-872-845 Supplier Chemglass Life Sciences Supplier No. CG1029G14
Adapter, Flow Control, Straight, 2mm Stpk, 14/20 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm and is supplied complete with a CG-423 retaining clip. Hose connection has an O.D. of 8mm at the middle serration.