Learn More
Chemglass Life Sciences Adapter, Flow Control, Straight, 2mm Stpk, 19/22 Inner Joint
SDP
Supplier:  Chemglass Life Sciences CG102919
Encompass_Preferred
Adapter, Flow Control, Straight, 2mm Stpk, 19/22 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration.
Catalog No. 50-872-844