Learn More
Chemglass Life Sciences Adapter, Flow Control, Minum-Ware
SDP
Catalog No. 501532678
Encompass_Preferred
Adapter, Flow Control, Minum-Ware. Used to regulate the input of liquids or gases into reactions. Stopcock is PTFE with a bore of 1mm. Inner joint is a 14/10.
Catalog No. 50-153-2678 Supplier Chemglass Life Sciences Supplier No. MW3001
Adapter, Flow Control, Minum-Ware. Used to regulate the input of liquids or gases into reactions. Stopcock is PTFE with a bore of 1mm. Inner joint is a 14/10.