Learn More
Chemglass Life Sciences Adapter, Flow Control, 90°, 2mm Stpk, 24/40 Outer Joint
SDP
Catalog No. CG1030A01
Encompass_Preferred
Adapter, Flow Control, 90¿, 2mm Stpk, 24/40 Outer Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration. With standard taper outer joint.
Catalog No. CG1030A01 Supplier Chemglass Life Sciences Supplier No. CG1030A01
Adapter, Flow Control, 90¿, 2mm Stpk, 24/40 Outer Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration. With standard taper outer joint.