Learn More
Chemglass Life Sciences Adapter, Flow Control, Straight, 2mm Stpk, 29/42 Inner Joint
SDP
Catalog No. CG102902
Encompass_Preferred
Adapter, Flow Control, Straight, 2mm Stpk, 29/42 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration.
Catalog No. CG102902 Supplier Chemglass Life Sciences Supplier No. CG102902
Adapter, Flow Control, Straight, 2mm Stpk, 29/42 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration.