Learn More
Chemglass Life Sciences Adapter, Flow Control, Chromatography, 75/50 Ball Joint
SDP
Catalog No. CG120111
Encompass_Preferred
Adapter, Flow Control, Chromatography, 75/50 Ball Joint. Flow control adapter for use with CG-1198 and CG-1199 chromatography columns along with CG-1200 reservoir. Adapter has a 0-4mm Chem-Cap metering plug.
Catalog No. CG120111 Supplier Chemglass Life Sciences Supplier No. CG120111
Adapter, Flow Control, Chromatography, 75/50 Ball Joint. Flow control adapter for use with CG-1198 and CG-1199 chromatography columns along with CG-1200 reservoir. Adapter has a 0-4mm Chem-Cap metering plug.