Learn More
Ace Glass, Inc. ADAPTER FOR -30 ADAPTER FOR -30
SDP
Catalog No. 50975465
Encompass_Preferred
Drying Tower Connecting Adapter for ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-465 Supplier Ace Glass, Inc. Supplier No. 781832
Drying Tower Connecting Adapter for ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.