Learn More
Ace Glass, Inc. ADAPTER FOR -34 ADAPTER FOR -34
SDP
Catalog No. 50979477
Encompass_Preferred
Catalyst Column Adapter for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-979-477 Supplier Ace Glass, Inc. Supplier No. 781836
Catalyst Column Adapter for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.