Learn More
MTC Bio Adapter for SureStrain™ Premium Cell Strainers, 25/PK
Catalog No. 501871045
Encompass
This item has a minimum qty of 3 per supplier requirements.
Reducing Adapter for 15mL/5mL tubes
Catalog No. 50-187-1045 Supplier MTC Bio Supplier No. C4015
This item has a minimum qty of 3 per supplier requirements.
Reducing Adapter for 15mL/5mL tubes