Learn More
Chemglass Life Sciences Adapter, Inlet Feed, Fritted, Complete, for Rotary Evaporator
SDP
Catalog No. CG132730
Encompass_Preferred
Inlet feed tube for “A” assemblies with a 19/38 joint, 90º bore stopcock and PTFE extension tube.
Catalog No. CG132730 Supplier Chemglass Life Sciences Supplier No. CG132730
Inlet feed tube for “A” assemblies with a 19/38 joint, 90º bore stopcock and PTFE extension tube.