Learn More
Chemglass Life Sciences Style "C" Gas Inlet Adapter, 45/50
SDP
Catalog No. 50873386
Encompass_Preferred
Gas inlet adapter with 45/50 inner joint, is used for connection of the gas source to the lower reservoir. Inlet connection is 1/2 Swagelok tubing for use with CG-1975-20 compression fitting. Adapter is available in 3 styles.Style "C": Adapter permits the gas to enter the flask above the sub-surface of the scrubbing fluid. When using this adapter, all of the gas to liquid interaction takes place in the column.
Catalog No. 50-873-386 Supplier Chemglass Life Sciences Supplier No. CG183020
Gas inlet adapter with 45/50 inner joint, is used for connection of the gas source to the lower reservoir. Inlet connection is 1/2 Swagelok tubing for use with CG-1975-20 compression fitting. Adapter is available in 3 styles.Style "C": Adapter permits the gas to enter the flask above the sub-surface of the scrubbing fluid. When using this adapter, all of the gas to liquid interaction takes place in the column.