Learn More
Chemglass Life Sciences Style "B" Gas Inlet Adapter, 45/50, (SUPPLIED WITH COMPLETE SCRUBBER)
SDP
Catalog No. 501531255
Encompass_Preferred
Style "B" Gas Inlet Adapter, 45/50, (SUPPLIED WITH COMPLETE SCRUBBER). Gas inlet adapter with 45/50 inner joint, is used for connection of the gas source to the lower reservoir. Inlet connection is 1/2 Swagelok tubing for use with CG-1975-20 compression fitting. Adapter has an extension tube without the fritted section to insert the gas into the scrubbing fluid sub-surface. PLEASE NOTE: THIS STYLE ADAPTER IS SUPPLIED WITH THE COMPLETE SCRUBBER.
Catalog No. 50-153-1255 Supplier Chemglass Life Sciences Supplier No. CG183019
Style "B" Gas Inlet Adapter, 45/50, (SUPPLIED WITH COMPLETE SCRUBBER). Gas inlet adapter with 45/50 inner joint, is used for connection of the gas source to the lower reservoir. Inlet connection is 1/2 Swagelok tubing for use with CG-1975-20 compression fitting. Adapter has an extension tube without the fritted section to insert the gas into the scrubbing fluid sub-surface. PLEASE NOTE: THIS STYLE ADAPTER IS SUPPLIED WITH THE COMPLETE SCRUBBER.