Learn More
Chemglass Life Sciences Glass Adapter Only, 29/42, Glass
SDP
Catalog No. CG131812
Encompass_Preferred
Glass Adapter Only, 29/42, Glass. Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator. Adapters have a top outer joint of the listed size and a 20-400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter.
Catalog No. CG131812 Supplier Chemglass Life Sciences Supplier No. CG131812
Glass Adapter Only, 29/42, Glass. Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator. Adapters have a top outer joint of the listed size and a 20-400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter.