Learn More
Fritsch Milling And Sizing Inc Adapter Grinding Bowls 125-150 Premium Line
SDP
Catalog No. 501534595
Encompass
Adapter for all grinding bowls for Planetary Mill PULVERISETTE 5 premium line 125-150 ml volume.
Essential, if only one grinding bowl is inserted in the grinding bowl holder.
Catalog No. 50-153-4595 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.6837.00
Adapter for all grinding bowls for Planetary Mill PULVERISETTE 5 premium line 125-150 ml volume.
Essential, if only one grinding bowl is inserted in the grinding bowl holder.