Learn More
Ace Glass, Inc. Adapter, O-ring grooved 3/8in tube with 0-8 stopcock valve
SDP
Catalog No. 50972234
Encompass_Preferred
Bottom outlet valve/adapter for use with big chromatography columns. Adapter has an O-ring grooved 3/8 tube and a 0-8mm threaded stopcock valve for controlling flow.
Catalog No. 50-972-234 Supplier Ace Glass, Inc. Supplier No. 583532
Bottom outlet valve/adapter for use with big chromatography columns. Adapter has an O-ring grooved 3/8 tube and a 0-8mm threaded stopcock valve for controlling flow.