Learn More
Ace Glass, Inc. Adapter, O-ring grooved 3/8in tube with straight tube for Swagelok
SDP
Catalog No. 50972235
Encompass_Preferred
Bottom outlet valve/adapter for use with big chromatography columns. Adapter has an O-ring grooved 3/8 tube and straight tube for use with Swagelok fittings.
Catalog No. 50-972-235 Supplier Ace Glass, Inc. Supplier No. 583534
Bottom outlet valve/adapter for use with big chromatography columns. Adapter has an O-ring grooved 3/8 tube and straight tube for use with Swagelok fittings.